3,5-Bis(trifluoromethyl)benzylamine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3104M-100G PC3104M-25G PC3104M-5G PC3104M-1G PC3104M-10G
APOSPC3104M-100GEA 199 EUR
APOSPC3104M-100G APOSPC3104M-25G APOSPC3104M-5G APOSPC3104M-1G APOSPC3104M-10G
3,5-Bis(trifluoromethyl)benzylamine 95%
3,5-Bis(trifluoromethyl)benzylamine
Formule: C₉H₇F₆N
Kookpunt: 82…84 °C/15mmHg
Smeltpunt: 50…55 °C
Vlampunt: 78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009909
CAS nummer: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR