Ethyl bromodifluoroacetate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3162-50G PC3162-250G PC3162-500G
APOSPC3162-50GEA 22 EUR
APOSPC3162-50G APOSPC3162-250G APOSPC3162-500G
Ethyl bromodifluoroacetate 98%
Ethyl bromodifluoroacetate

Treatment of the Reformatsky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. Please enquire for references and other data for this item.

Formule: BrF₂CCOOCH₂CH₃
Kookpunt: 112 °C/700mmHg
Dichtheid: 1,583 g/cm³ (25 °C)
Vlampunt: 21 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042069
CAS nummer: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR