α,3-Dibromo-2-fluorotoluene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3168-1G PC3168-5G PC3168-25G
APOSPC3168-1GEA 22 EUR
APOSPC3168-1G APOSPC3168-5G APOSPC3168-25G
α,3-Dibromo-2-fluorotoluene
α,3-Dibromo-2-fluorotoluene
Formule: C₇H₅Br₂F
Kookpunt: 60 °C/0,1mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09842442
CAS nummer: 149947-16-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR