8-Fluoroquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3205-1G PC3205-25G PC3205-5G
APOSPC3205-1GEA 22 EUR
APOSPC3205-1G APOSPC3205-25G APOSPC3205-5G
8-Fluoroquinoline
8-Fluoroquinoline
Formule: C₉H₆FN
Kookpunt: 148…150 °C/30mmHg
Dichtheid: 1,215 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01685514
CAS nummer: 394-68-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR