3-(Trifluoromethyl)phenyl isothiocyanate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: α,α,α-Trifluoro-m-tolyl isothiocyanate

PC32284-5G PC32284-25G
APOSPC32284-5GEA 58.4 EUR
APOSPC32284-5G APOSPC32284-25G
3-(Trifluoromethyl)phenyl isothiocyanate 98%
3-(Trifluoromethyl)phenyl isothiocyanate
Formule: C₈H₄F₃NS
MW: 203.19 g/mol
Kookpunt: 206…208 °C
Dichtheid: 1.335 g/cm³ (25 °C)
Vlampunt: 94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004806
CAS nummer: 1840-19-3
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR