2,4-Difluorothiobenzamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC32594-5G PC32594-250MG PC32594-100MG
APOSPC32594-5GEA 263 EUR
APOSPC32594-5G APOSPC32594-250MG APOSPC32594-100MG
2,4-Difluorothiobenzamide 95%
2,4-Difluorothiobenzamide
Formule: C₇H₅F₂NS
Smeltpunt: 128…132 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085011
CAS nummer: 175276-92-3
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR