2-Nitro-6-(trifluoromethyl)benzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC32747-1G PC32747-5G PC32747-25G
APOSPC32747-1GEA 22 EUR
APOSPC32747-1G APOSPC32747-5G APOSPC32747-25G
2-Nitro-6-(trifluoromethyl)benzonitrile
2-Nitro-6-(trifluoromethyl)benzonitrile
Formule: C₈H₃F₃N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16606330
CAS nummer: 1227581-80-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR