3-(4-Fluorophenyl)pyrazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3297-1G PC3297-5G PC3297-25G
APOSPC3297-1GEA 22 EUR
APOSPC3297-1G APOSPC3297-5G APOSPC3297-25G
3-(4-Fluorophenyl)pyrazole
3-(4-Fluorophenyl)pyrazole
Formule: C₉H₇FN₂
Smeltpunt: 101…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00665857
CAS nummer: 154258-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR