4-Fluoro-2-iodoaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3452-100G PC3452-25G PC3452-5G
APOSPC3452-100GEA 103 EUR
APOSPC3452-100G APOSPC3452-25G APOSPC3452-5G
4-Fluoro-2-iodoaniline 98%
4-Fluoro-2-iodoaniline
Formule: C₆H₅FIN
Kookpunt: 74…76 °C
Dichtheid: 1,99 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042584
CAS nummer: 61272-76-2
UN: 2810
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR