2,3-Dichloro-6-fluorophenacyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3541-10G PC3541-5G
APOSPC3541-10GEA 448 EUR
APOSPC3541-10G APOSPC3541-5G
2,3-Dichloro-6-fluorophenacyl bromide
2,3-Dichloro-6-fluorophenacyl bromide
Formule: C₈H₄BrCl₂FO
Smeltpunt: 48…50 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07368755
CAS nummer: 886762-53-4
UN: 1759
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR