α-Chloro-3-fluorotoulene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride

PC3710-100G PC3710-1KG PC3710-500G
APOSPC3710-100GEA 74.6 EUR
APOSPC3710-100G APOSPC3710-1KG APOSPC3710-500G
α-Chloro-3-fluorotoulene 97%
α-Chloro-3-fluorotoulene
Formule: C₇H₆ClF
MW: 144.58 g/mol
Kookpunt: 84 °C/29mmHg
Dichtheid: 1.213 g/cm³ (25 °C)
Vlampunt: 58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000904
CAS nummer: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR