5-Fluorocytosine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3735-100G PC3735-25G PC3735-250G
APOSPC3735-100GEA 35.1 EUR
APOSPC3735-100G APOSPC3735-25G APOSPC3735-250G
5-Fluorocytosine 98%
5-Fluorocytosine
Formule: C₄H₄FN₃O
Smeltpunt: 298…300 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006035
CAS nummer: 2022-85-7
EINECS: 217-968-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR