tert-Butyl pentafluoropropionate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC3772-25G PC3772-5G
APOSPC3772-25GEA 667 EUR
APOSPC3772-25G APOSPC3772-5G
tert-Butyl pentafluoropropionate 98%
tert-Butyl pentafluoropropionate
Formule: C₇H₉F₅O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00464380
CAS nummer: 55258-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR