1-Fluoro-4-iodobenzene 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3780-100G PC3780-500G PC3780-50G
APOSPC3780-100GEA 36.6 EUR
APOSPC3780-100G APOSPC3780-500G APOSPC3780-50G
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene

Tet. Lett., 33 7499 (1992): Using LDA regioselective metallation can be achieved ortho- to the fluorine atom.J.Med.Chem,40,717(1997):5-iodoanthranilic acid & deriv.

Formule: C₆H₄FI
Kookpunt: 182…184 °C
Smeltpunt: –20 °C
Dichtheid: 1,925 g/cm³ (25 °C)
Vlampunt: 68 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001052
CAS nummer: 352-34-1
EINECS: 206-522-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR