1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC37883-250MG PC37883-1G PC37883-5G
APOSPC37883-250MGEA 191 EUR
APOSPC37883-250MG APOSPC37883-1G APOSPC37883-5G
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone 95%
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone
Formule: C₈H₅BrClFO
MDL Nummer: MFCD26679371
CAS nummer: 1536939-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR