4-Fluoro-2-iodotoluene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3810-100G PC3810-25G
APOSPC3810-100GEA 116 EUR
APOSPC3810-100G APOSPC3810-25G
4-Fluoro-2-iodotoluene 98%
4-Fluoro-2-iodotoluene
Formule: C₇H₆FI
Kookpunt: 204…208 °C
Dichtheid: 1,81 g/cm³ (25 °C)
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013709
CAS nummer: 13194-67-7
EINECS: 236-153-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR