4-Fluoro-m-toluidine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3820-5G PC3820-100G PC3820-25G
APOSPC3820-5GEA 14.8 EUR
APOSPC3820-5G APOSPC3820-100G APOSPC3820-25G
4-Fluoro-m-toluidine 98%
4-Fluoro-m-toluidine
Formule: C₇H₈FN
Kookpunt: 85…86 °C/9mmHg
Smeltpunt: 32…38 °C
Dichtheid: 1,12 g/cm³ (25 °C)
Vlampunt: 95 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00025294
CAS nummer: 452-69-7
EINECS: 207-207-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR