2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3823R-50MG PC3823R-1G PC3823R-5G
APOSPC3823R-50MGEA 22 EUR
APOSPC3823R-50MG APOSPC3823R-1G APOSPC3823R-5G
2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%
2-Fluoro-6-(4-methylphenoxy)benzonitrile
Formule: C₁₄H₁₀FNO
Smeltpunt: 93…95 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00068203
CAS nummer: 175204-08-7
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR