2-Fluoro-6-(4-methylphenylthio)benzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3823X-1G
APOSPC3823X-1GEA 42.4 EUR
APOSPC3823X-1G
2-Fluoro-6-(4-methylphenylthio)benzonitrile 97%
2-Fluoro-6-(4-methylphenylthio)benzonitrile
Formule: C₁₄H₁₀FNS
Smeltpunt: 92…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00068207
CAS nummer: 175204-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR