5-(2-Fluorophenyl)-1H-tetrazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3845-1G PC3845-250MG
APOSPC3845-1GEA 29.2 EUR
APOSPC3845-1G APOSPC3845-250MG
5-(2-Fluorophenyl)-1H-tetrazole
5-(2-Fluorophenyl)-1H-tetrazole
Formule: C₇H₅FN₄
Smeltpunt: 157…159 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00798373
CAS nummer: 50907-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR