4-Fluoro-3-nitroaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3880-100G PC3880-25G PC3880-500G
APOSPC3880-100GEA 24.9 EUR
APOSPC3880-100G APOSPC3880-25G APOSPC3880-500G
4-Fluoro-3-nitroaniline 98%
4-Fluoro-3-nitroaniline
Formule: C₆H₅FN₂O₂
MW: 156.12 g/mol
Smeltpunt: 94…98 °C
Vlampunt: 91 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007833
CAS nummer: 364-76-1
EINECS: 206-665-5
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR