5-Chloro-2,4-difluorobenzenesulphonamide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC3926-1G PC3926-5G
APOSPC3926-1GEA 110 EUR
APOSPC3926-1G APOSPC3926-5G
5-Chloro-2,4-difluorobenzenesulphonamide 97%
5-Chloro-2,4-difluorobenzenesulphonamide
Formule: C₆H₄ClF₂NO₂S
Smeltpunt: 118…120 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01940427
CAS nummer: 13656-56-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR