trans-4,4'-Difluorochalcone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC3949-100G PC3949-25G PC3949-5G PC3949-50G
APOSPC3949-100GEA 243 EUR
APOSPC3949-100G APOSPC3949-25G APOSPC3949-5G APOSPC3949-50G
trans-4,4'-Difluorochalcone 98%
trans-4,4'-Difluorochalcone
Formule: C₁₅H₁₀F₂O
Smeltpunt: 117…119 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00219906
CAS nummer: 102692-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR