5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC400031-1G PC400031-5G
APOSPC400031-1GEA 76 EUR
APOSPC400031-1G APOSPC400031-5G
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
Formule: C₁₃H₁₉FO₂Si
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD17676576
CAS nummer: 113984-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR