4-(Difluoromethoxy)pyridine-2-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC400110-100MG PC400110-250MG
APOSPC400110-100MGEA 424 EUR
APOSPC400110-100MG APOSPC400110-250MG
4-(Difluoromethoxy)pyridine-2-carboxylic acid
4-(Difluoromethoxy)pyridine-2-carboxylic acid
Formule: C₇H₅F₂NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD27995942
CAS nummer: 1707365-38-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR