2,3-Difluoro-4-(trifluoromethyl)aniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4002-1G PC4002-5G
APOSPC4002-1GEA 132 EUR
APOSPC4002-1G APOSPC4002-5G
2,3-Difluoro-4-(trifluoromethyl)aniline 98%
2,3-Difluoro-4-(trifluoromethyl)aniline
Formule: C₇H₄F₅N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236206
CAS nummer: 123950-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR