6-(Trifluoromethyl)pyrimidin-4-amine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC400532-1G
APOSPC400532-1GEA 51.2 EUR
APOSPC400532-1G
6-(Trifluoromethyl)pyrimidin-4-amine
6-(Trifluoromethyl)pyrimidin-4-amine
Formule: C5H4F3N3
Smeltpunt: 165…170 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08436657
CAS nummer: 672-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR