4-Nitro-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC400600-1G
APOSPC400600-1GEA 520 EUR
APOSPC400600-1G
4-Nitro-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid
4-Nitro-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid
Formule: C₅H₂F₃N₃O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11845715
CAS nummer: 1322805-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR