4-(4-Trifluoromethylpyridin-2-yl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC400664-1G PC400664-250MG PC400664-500MG
APOSPC400664-1GEA 297 EUR
APOSPC400664-1G APOSPC400664-250MG APOSPC400664-500MG
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
Formule: C₁₃H₈F₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18384864
CAS nummer: 1299607-71-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR