Methyl-3-fluoro-2-nitrobenzoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC400724-1G PC400724-25G PC400724-5G
APOSPC400724-1GEA 51.2 EUR
APOSPC400724-1G APOSPC400724-25G APOSPC400724-5G
Methyl-3-fluoro-2-nitrobenzoate
Methyl-3-fluoro-2-nitrobenzoate
Formule: C₈H₆FNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11520393
CAS nummer: 1214353-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR