Fmoc-4-(trifluoromethyl)-D-phenylalanine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC4013-1G PC4013-5G
APOSPC4013-1GEA 122 EUR
APOSPC4013-1G APOSPC4013-5G
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Formule: C₂₅H₂₀F₃NO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00797582
CAS nummer: 238742-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR