4-Bromo-3-chloro-5,8-difluoroquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC404606-1G
APOSPC404606-1GEA 628 EUR
APOSPC404606-1G
4-Bromo-3-chloro-5,8-difluoroquinoline
4-Bromo-3-chloro-5,8-difluoroquinoline
Formule: C₉H₃BrClF₂N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13619736
CAS nummer: 1210275-42-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR