5-Nitro-2-(trifluoromethoxy)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC408278-10G PC408278-1G PC408278-5G
APOSPC408278-10GEA 622 EUR
APOSPC408278-10G APOSPC408278-1G APOSPC408278-5G
5-Nitro-2-(trifluoromethoxy)benzoic acid
5-Nitro-2-(trifluoromethoxy)benzoic acid
Formule: C₈H₄F₃NO₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11847469
CAS nummer: 307989-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR