1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC410439-1G
APOSPC410439-1GEA 651 EUR
APOSPC410439-1G
1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole
1-(2,2-Difluoroethyl)-4-nitro-1H-pyrazole
Formule: C₅H₅F₂N₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08556227
CAS nummer: 1006442-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR