4-(3,5-Difluorophenoxy)phenylboronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC412134-1G PC412134-250MG
APOSPC412134-1GEA 456 EUR
APOSPC412134-1G APOSPC412134-250MG
4-(3,5-Difluorophenoxy)phenylboronic acid
4-(3,5-Difluorophenoxy)phenylboronic acid
Formule: C₁₂H₉BF₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD14636486
CAS nummer: 1029438-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR