2-Fluorophenylhydrazinium chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4152-100G PC4152-25G PC4152-5G
APOSPC4152-100GEA 76 EUR
APOSPC4152-100G APOSPC4152-25G APOSPC4152-5G
2-Fluorophenylhydrazinium chloride 98%
2-Fluorophenylhydrazinium chloride
Formule: C₆H₈ClFN₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012927
CAS nummer: 2924-15-4
EINECS: 220-885-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR