4-Fluoro-α-methylbenzyl alcohol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4173-1G
APOSPC4173-1GEA 22 EUR
APOSPC4173-1G
4-Fluoro-α-methylbenzyl alcohol 97%
4-Fluoro-α-methylbenzyl alcohol
Formule: C₈H₉FO
Kookpunt: 91 °C/10mmHg
Dichtheid: 1,109 g/cm³ (25 °C)
Vlampunt: 91 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004515
CAS nummer: 403-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR