1-(2-Fluorophenyl)-2-thiourea 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC4175M-25G PC4175M-5G
APOSPC4175M-25GEA 70.1 EUR
APOSPC4175M-25G APOSPC4175M-5G
1-(2-Fluorophenyl)-2-thiourea 97%
1-(2-Fluorophenyl)-2-thiourea
Formule: (C₆H₄F)NC(S)NH₂
Smeltpunt: 143…145 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041148
CAS nummer: 656-32-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR