N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC4175W-1G PC4175W-5G
APOSPC4175W-1GEA 92.6 EUR
APOSPC4175W-1G APOSPC4175W-5G
N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide 98%
N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide
Formule: C₁₄H₉F₄NO
Smeltpunt: 101…102 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00045089
CAS nummer: 33489-71-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR