2,6-Dichloro-4-fluoroiodobenzene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC421169-250MG PC421169-1G
APOSPC421169-250MGEA 95 EUR
APOSPC421169-250MG APOSPC421169-1G
2,6-Dichloro-4-fluoroiodobenzene 95%
2,6-Dichloro-4-fluoroiodobenzene
Formule: C₆H₂Cl₂FI
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13194304
CAS nummer: 939990-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR