4-Cyclopropyl-2-fluoro-6-methylbenzamide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC421217-100MG PC421217-1G PC421217-250MG
APOSPC421217-100MGEA 453 EUR
APOSPC421217-100MG APOSPC421217-1G APOSPC421217-250MG
4-Cyclopropyl-2-fluoro-6-methylbenzamide 95%
4-Cyclopropyl-2-fluoro-6-methylbenzamide
Formule: C₁₁H₁₂FNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD20726776
CAS nummer: 1242156-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR