4-Fluoro-3-(trifluoromethyl)benzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4372Q-100G PC4372Q-25G PC4372Q-5G
APOSPC4372Q-100GEA 135 EUR
APOSPC4372Q-100G APOSPC4372Q-25G APOSPC4372Q-5G
4-Fluoro-3-(trifluoromethyl)benzaldehyde 97%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Formule: C₈H₄F₄O
Kookpunt: 60 °C/0,75mmHg
Dichtheid: 1,408 g/cm³ (25 °C)
Vlampunt: 80 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061314
CAS nummer: 67515-60-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR