4-Fluoro-3-(trifluoromethyl)benzylamine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC4374Z-25G PC4374Z-5G
APOSPC4374Z-25GEA 342 EUR
APOSPC4374Z-25G APOSPC4374Z-5G
4-Fluoro-3-(trifluoromethyl)benzylamine 97%
4-Fluoro-3-(trifluoromethyl)benzylamine
Formule: C₈H₇F₄N
Kookpunt: 101 °C/3mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070814
CAS nummer: 67515-74-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR