3-Fluoro-DL-tyrosine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4379-500MG PC4379-1G PC4379-250MG
APOSPC4379-500MGEA 0 EUR
APOSPC4379-500MG APOSPC4379-1G APOSPC4379-250MG
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine
Formule: C₉H₁₀FNO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063075
CAS nummer: 403-90-7
EINECS: 206-964-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR