5-Fluoro-2-nitropyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC445034-250MG PC445034-1G PC445034-5G
APOSPC445034-250MGEA 43.8 EUR
APOSPC445034-250MG APOSPC445034-1G APOSPC445034-5G
5-Fluoro-2-nitropyridine
5-Fluoro-2-nitropyridine
Formule: C5H3FN2O2
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03092917
CAS nummer: 779345-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR