N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC446004-1G PC446004-500MG
APOSPC446004-1GEA 184 EUR
APOSPC446004-1G APOSPC446004-500MG
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
N'-[1-Amino-1-(3,5-bis-(trifluoromethyl)phenyl)methylidene]hydrazine carboxylic acid-tert-butyl ester
Formule: C₁₄H₁₅F₆N₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10568191
CAS nummer: 1053655-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR