2-Bromo-3',5'-difluoroacetophenone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC4473-1G PC4473-5G
APOSPC4473-1GEA 103 EUR
APOSPC4473-1G APOSPC4473-5G
2-Bromo-3',5'-difluoroacetophenone 98%
2-Bromo-3',5'-difluoroacetophenone
Formule: C₈H₅BrF₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094245
CAS nummer: 220607-75-0
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR