1-(Oxan-2-yl)-3-(trifluoromethyl)pyrazole-5-carboxaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC448029-1G
APOSPC448029-1GEA 291 EUR
APOSPC448029-1G
1-(Oxan-2-yl)-3-(trifluoromethyl)pyrazole-5-carboxaldehyde
1-(Oxan-2-yl)-3-(trifluoromethyl)pyrazole-5-carboxaldehyde
Formule: C₁₀H₁₁F₃N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD24448787
CAS nummer: 1437794-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR