2-(1,1,2,2-Tetrafluoro-2-phenylethyl)pyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC450020-100MG PC450020-250MG
APOSPC450020-100MGEA 199 EUR
APOSPC450020-100MG APOSPC450020-250MG
2-(1,1,2,2-Tetrafluoro-2-phenylethyl)pyridine
2-(1,1,2,2-Tetrafluoro-2-phenylethyl)pyridine
Formule: C₁₃H₉F₄N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22377940
CAS nummer: 1378261-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR