Sodium dichlorofluoroacetate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC450243-1G PC450243-5G
APOSPC450243-1GEA 65.8 EUR
APOSPC450243-1G APOSPC450243-5G
Sodium dichlorofluoroacetate 98%
Sodium dichlorofluoroacetate
Formule: C₂Cl₂FO₂·Na
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02696369
CAS nummer: 2837-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR